Invitrogen
Manufacturer Code:PA5105253
Catalog # PIPA5105253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NAKED2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2210403L10Rik AW212591 Dvl-binding protein NKD2 hNkd2 mNkd2 naked cuticle 1 homolog naked cuticle 2 homolog naked cuticle 2 homolog (Drosophila) naked cuticle homolog 2 naked cuticle homolog 2 (Drosophila) naked cuticle-2 Naked2 naked-2 NKD2 protein naked cuticle homolog 2; Dvl-binding protein NKD2; mNkd2; naked cuticle 1 homolog; naked cuticle 2 homolog; Naked-2; Protein naked cuticle homolog 2
Gene Aliases: 2210403L10Rik; AW212591; Naked2; NKD2
UniProt ID: (Human) Q969F2, (Mouse) Q8VE28
Entrez Gene ID: (Human) 85409, (Rat) 308068, (Mouse) 72293
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.