Invitrogen
Manufacturer Code:PA527556
Catalog # PIPA527556
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 358 and 615 of Human NYREN18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BS4 NUB1L NYREN18 NEDD8 ultimate buster 1 NEDD8 ultimate buster-1 renal carcinoma antigen NY-REN-18; NEDD8 ultimate buster 1; NEDD8 ultimate buster-1; Negative regulator of ubiquitin-like proteins 1; Renal carcinoma antigen NY-REN-18
Gene Aliases: BS4; NUB1; NUB1L; NYREN18
UniProt ID: (Human) Q9Y5A7
Entrez Gene ID: (Human) 51667
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.