Invitrogen
Manufacturer Code:PA512190
Catalog # PIPA512190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 87-117 amino acids from the N-terminal region of human NYREN18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NEDD8 ultimate buster 1; NEDD8 ultimate buster-1; NEDD8 ultimate buster-1 NUB1 Negative regulator of ubiquitin-like proteins 1 Renal carcinoma antigen NY-REN-18; Negative regulator of ubiquitin-like proteins 1; Protein BS4; Renal carcinoma antigen NY-REN-18
Gene Aliases: 4931404D21Rik; 6330412F12Rik; BS4; NUB1; NUB1L; NY-REN-18; NYREN18
UniProt ID: (Human) Q9Y5A7, (Mouse) P54729
Entrez Gene ID: (Human) 51667, (Mouse) 53312
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.