Proteintech
Manufacturer Code:251081AP150UL
Catalog # 50-173-4726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NYAP2 Fusion Protein Ag19004 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1486 KIAA1486 protein NYAP2; neuronal tyrosine phosphorylated adaptor for phosphoinositide 3-kinase adaptor 2; Neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 2
Gene Aliases: 9430031J16Rik; Jr6; KIAA1486; NYAP2
UniProt ID: (Human) Q9P242
Entrez Gene ID: (Human) 57624, (Mouse) 241134
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.