Novus Biologicals
Manufacturer Code:NBP293010100UL
Catalog # NB027915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 110-180 of human NXT2 (NP_061168.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM-025 NTF2-related export protein 2 nuclear transport factor 2-like export factor 2 protein p15-2; NTF2-related export protein 2; nuclear transport factor 2-like export factor 2; Protein p15-2
Gene Aliases: BM-025; DC9; NXT2; P15-2
UniProt ID: (Human) Q9NPJ8
Entrez Gene ID: (Human) 55916
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.