Invitrogen
Manufacturer Code:PA565471
Catalog # PIPA565471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human NXT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MTR2 P15 NTF2-related export protein 1 NTX2-like export factor1 NUTF-like export factor 1 protein p15; NTF2-like export factor 1; NTF2-related export protein 1; NTX2-like export factor1; nuclear transport factor 2-like export factor 1; NUTF-like export factor 1; Protein p15
Gene Aliases: MTR2; NXT1; P15
UniProt ID: (Human) Q9UKK6
Entrez Gene ID: (Human) 29107
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.