Novus Biologicals
Manufacturer Code:NBP179561
Catalog # NBP179561
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal of human NXT1The immunogen for this antibody is NXT1. Peptide sequence GTATLVWNGNAVSGQESLSEFFEMLPSSEFQISVVDCQPVHDEATPSQTT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MTR2NTF2-related export protein 1 NTF2-like export factor 1 NTX2-like export factor1 NUTF-like export factor 1 P15 Protein p15; NTF2-like export factor 1; NTF2-related export protein 1; NTX2-like export factor1; nuclear transport factor 2-like export factor 1; NUTF-like export factor 1; Protein p15
Gene Aliases: MTR2; NXT1; P15
UniProt ID: (Human) Q9UKK6
Entrez Gene ID: (Human) 29107
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.