Invitrogen
Manufacturer Code:PA536496
Catalog # PIPA536496
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 221-270 of human Neurexophilin-4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternative protein NXPH4; Neurexophilin 4 NXPH4 NPH4 UNQ1928/PRO4403; Neurexophilin-4
Gene Aliases: 1110036M10Rik; AI851051; NPH4; NXPH4; UNQ1928/PRO4403
UniProt ID: (Human) O95158, (Rat) Q9Z2N4
Entrez Gene ID: (Human) 11247, (Mouse) 104080, (Rat) 59316
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.