Novus Biologicals
Manufacturer Code:NBP157773
Catalog # NBP157773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to NXPH4 (neurexophilin 4) The peptide sequence was selected from the N terminal of NXPH4)(50ug). Peptide sequence MRLLPEWFLLLFGPWLLRKAVSAQIPESGRPQYLGLRPAAAGAGAPGQQL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternative protein NXPH4; neurexophilin 4 NPH4neurexophilin-4; Neurexophilin-4
Gene Aliases: NPH4; NXPH4; UNQ1928/PRO4403
UniProt ID: (Human) O95158
Entrez Gene ID: (Human) 11247
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.