Invitrogen
Manufacturer Code:PA539289
Catalog # PIPA539289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human NXPH3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1159 NPH3; Neurexophilin-3
Gene Aliases: KIAA1159; NPH3; NXPH3; UNQ687/PRO1327
UniProt ID: (Human) O95157, (Mouse) Q91VX5, (Rat) Q9Z2N5
Entrez Gene ID: (Human) 11248, (Mouse) 104079, (Rat) 59315
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.