Invitrogen
Manufacturer Code:PA575469
Catalog # PIPA575469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 80-130 of Human NXPH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neurexophilin-1; Neurexophilin-1 NXPH1 NPH1 Nbla00697; Neurophilin
Gene Aliases: C130005L03Rik; Nbla00697; NPH1; NXPH1
UniProt ID: (Human) P58417, (Mouse) Q61200, (Rat) Q63366
Entrez Gene ID: (Human) 30010, (Mouse) 18231, (Rat) 25501
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.