Novus Biologicals
Manufacturer Code:NBP273134B
Catalog # NBP273134B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B11 |
Immunogen | Full length recombinant protein of human NXNL2 (NP_660326) produced in HEK293T cells. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: C9orf121 chromosome 9 open reading frame 121 nucleoredoxin-like 2 nucleoredoxin-like protein 2 RDCVF2; Nucleoredoxin-like protein 2; RdCVF2; Rod-derived cone viability factor 2
Gene Aliases: C9orf121; NXNL2; RDCVF2
UniProt ID: (Human) Q5VZ03
Entrez Gene ID: (Human) 158046
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.