Invitrogen
Manufacturer Code:PA524065
Catalog # PIPA524065
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 80-108 amino acids from the central region of human NXNL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C9orf121 RDCVF2 nucleoredoxin-like protein 2; Nucleoredoxin-like protein 2; RdCVF2; Rod-derived cone viability factor 2
Gene Aliases: 4930519N16Rik; C9orf121; NXNL2; RDCVF2
UniProt ID: (Human) Q5VZ03, (Mouse) Q9D531
Entrez Gene ID: (Human) 158046, (Mouse) 75124
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.