Invitrogen
Manufacturer Code:PA5100297
Catalog # PIPA5100297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NXN corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gn25 l11Jus13 NRX Nucleoredoxin nucleoredoxin 1 Nxn Protein Red-1 TRG-4; Nucleoredoxin; nucleoredoxin 1; Protein Red-1
Gene Aliases: Gn25; l11Jus13; NRX; NXN; TRG-4
UniProt ID: (Human) Q6DKJ4, (Mouse) P97346
Entrez Gene ID: (Human) 64359, (Mouse) 18230, (Rat) 360577
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.