Novus Biologicals
Manufacturer Code:NBP157260
Catalog # NBP157260
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to NXF5 (nuclear RNA export factor 5) The peptide sequence was selected from the middle region of NXF5. Peptide sequence ITERNFPELLSLNLCNNKLYQLDGLSDITEKAPKVKTLNLSKNKLESAWE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nuclear RNA export factor 5; nuclear RNA export factor 5 TAPL1 TAPL-1 TAP-like protein 1; TAP-like protein 1; TAPL-1
Gene Aliases: NXF5; TAPL1
UniProt ID: (Human) Q9H1B4
Entrez Gene ID: (Human) 55998
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.