Invitrogen
Manufacturer Code:PA542841
Catalog # PIPA542841
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human NUSAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nucleolar and spindle-associated protein 1; nucleolar protein ANKT; NuSAP
Gene Aliases: 2610201A12Rik; AI481307; ANKT; AW547774; BB165529; BM-037; BM037; fb76a01; fi37a11; LNP; NUSAP; NUSAP1; PRO0310; PRO0310p1; Q0310; SAPL; sb:cb490; wu:fb76a01; wu:fi37a11; YF-9
UniProt ID: (Bovine) Q2YDJ0, (Human) Q9BXS6, (Mouse) Q9ERH4, (Zebrafish) A1L2F3
Entrez Gene ID: (Bovine) 616028, (Dog) 475154, (Rabbit) 100349779, (Human) 51203, (Horse) 100071174, (Rat) 311336, (Mouse) 108907, (Guinea pig) 100732320, (Zebrafish) 567446
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.