Invitrogen
Manufacturer Code:PA5111726
Catalog # PIPA5111726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human NUSAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610201A12Rik AI481307 ANKT AW547774 BB165529 BM037 BM-037 BOS_10744 fb76a01 fi37a11 LNP Nucleolar and spindle-associated protein 1 NuSAP nusap1 PRO0310 PRO0310p1 Q0310 SAPL sb:cb490 wu:fb76a01 wu:fi37a11 YF-9; Nucleolar and spindle-associated protein 1; nucleolar protein ANKT; NuSAP
Gene Aliases: ANKT; BM-037; BM037; LNP; NUSAP; NUSAP1; PRO0310; PRO0310p1; Q0310; SAPL
UniProt ID: (Human) Q9BXS6
Entrez Gene ID: (Human) 51203
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.