Abnova
Manufacturer Code:PAB6588
Catalog # 89-132-516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human NUPR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of NUPR1.
For Research Use Only
Protein Aliases: Candidate of metastasis 1; COM1; Nuclear protein 1; nuclear protein, transcriptional regulator, 1; nuclear transcriptional regulator protein 1; P8; Protein p8
Gene Aliases: COM1; NUPR1; P8
UniProt ID: (Human) O60356
Entrez Gene ID: (Human) 26471
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.