Proteintech
Manufacturer Code:150561AP150UL
Catalog # 50-109-2790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NUPR1 Fusion Protein Ag7039 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Candidate of metastasis 1; Candidate of metastasis 1 COM1 nuclear protein 1 NUPR1 P8 Protein p8; Nuclear protein 1; nuclear protein, transcriptional regulator, 1; nuclear transcriptional regulator protein 1; Protein p8
Gene Aliases: COM1; NUPR1; P8
UniProt ID: (Human) O60356
Entrez Gene ID: (Human) 26471
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.