Invitrogen
Manufacturer Code:PA5113478
Catalog # PIPA5113478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Nucleoporin p58/p45 protein (211-480AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700017F11Rik 58 kDa nucleoporin KIAA0410 mKIAA0410 nucleoporin 58 nucleoporin 58kDa nucleoporin like 1 nucleoporin p45 nucleoporin p58 Nucleoporin p58/p45 nucleoporin-like 1 Nucleoporin-like protein 1 NUP45 NUP58 NUPL1 p58/p45 PRO2463; 58 kDa nucleoporin; nucleoporin 58kDa; nucleoporin like 1; Nucleoporin p58/p45; Nucleoporin-like protein 1
Gene Aliases: 1700017F11Rik; KIAA0410; mKIAA0410; NUP45; NUP58; NUPL1; PRO2463
UniProt ID: (Human) Q9BVL2, (Mouse) Q8R332
Entrez Gene ID: (Human) 9818, (Mouse) 71844
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.