Novus Biologicals
Manufacturer Code:NBP29428320UL
Catalog # NB023122
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of Human NUP58. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 58 kDa nucleoporin; KIAA0410PRO2463 nucleoporin like 1 nucleoporin p58/p45 Nucleoporin-like protein 1; nucleoporin 58kDa; nucleoporin like 1; Nucleoporin p58/p45; Nucleoporin-like protein 1
Gene Aliases: KIAA0410; NUP45; NUP58; NUPL1; PRO2463
UniProt ID: (Human) Q9BVL2
Entrez Gene ID: (Human) 9818
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.