Proteintech
Manufacturer Code:199071AP150UL
Catalog # 50-173-2594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NUPL1 Fusion Protein Ag13901 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 58 kDa nucleoporin; KIAA0410 nucleoporin like 1 Nucleoporin like protein 1 Nucleoporin p58/p45 NUPL1 PRO2463; nucleoporin 58kDa; nucleoporin like 1; nucleoporin p45; nucleoporin p58; Nucleoporin p58/p45; nucleoporin-like 1; Nucleoporin-like protein 1
Gene Aliases: 1700017F11Rik; KIAA0410; mKIAA0410; NUP45; NUP58; NUPL1; p58/p45; PRO2463
UniProt ID: (Human) Q9BVL2, (Mouse) Q8R332, (Rat) P70581
Entrez Gene ID: (Human) 9818, (Mouse) 71844, (Rat) 245922
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.