Proteintech
Manufacturer Code:123291AP150UL
Catalog # 50-172-7898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NUP98-NUP96 Fusion Protein Ag3016 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 96 kDa nucleoporin; 96 kDa nucleoporin ADAR2 ADIR2 Nucleoporin Nup96 NUP196 NUP96 NUP98 NUP96 NUP98-NUP96; 98 kDa nucleoporin; GLFG-repeat containing nucleoporin; Nuclear pore complex protein Nup96; Nuclear pore complex protein Nup98; Nuclear pore complex protein Nup98-Nup96; nucleoporin 98kD; nucleoporin 98kDa; Nucleoporin Nup96; Nucleoporin Nup98; Nup96; Nup98; Nup98-Nup96; NUP98/PHF23 fusion 2 protein
Gene Aliases: 4732457F17; ADAR2; ADIR2; AI849286; NUP196; NUP96; NUP98
UniProt ID: (Human) P52948, (Rat) P49793, (Mouse) Q6PFD9
Entrez Gene ID: (Human) 4928, (Rat) 81738, (Mouse) 269966
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.