Invitrogen
Manufacturer Code:PA517406
Catalog # PIPA517406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding Leu205 of human NUP98 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 96 kDa nucleoporin; 98 kDa nucleoporin; GLFG-repeat containing nucleoporin; GLFG-repeat containing nucleoporin Nup98-Nup96 OTTHUMP00000013954 nuclear pore complex protein Nup98-Nup96; Nuclear pore complex protein Nup96; Nuclear pore complex protein Nup98; Nuclear pore complex protein Nup98-Nup96; nucleoporin 98kD; nucleoporin 98kDa; Nucleoporin Nup96; Nucleoporin Nup98; Nup96; Nup98; Nup98-Nup96; NUP98/PHF23 fusion 2 protein
Gene Aliases: 4732457F17; ADAR2; ADIR2; AI849286; NUP196; NUP96; NUP98
UniProt ID: (Human) P52948, (Rat) P49793, (Mouse) Q6PFD9
Entrez Gene ID: (Human) 4928, (Dog) 476822, (Rat) 81738, (Mouse) 269966, (Xenopus) 446233
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.