Invitrogen
Manufacturer Code:PA5115680
Catalog # PIPA5115680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NUP98 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4732457F17 96 kDa nucleoporin 98 kDa nucleoporin ADAR2 ADIR2 AI849286 GLFG-repeat containing nucleoporin Nuclear pore complex protein Nup96 Nuclear pore complex protein Nup98 nuclear pore complex protein Nup98 nuclear pore complex protein Nup98-Nup96 nuclear pore complex protein Nup98-Nup96 nucleoporin 98 nucleoporin 98 and 96 precursor nucleoporin 98 L homeolog nucleoporin 98kD nucleoporin 98kDa Nucleoporin Nup96 Nucleoporin Nup98 NUP196 nup96 NUP98 nup98.L NUP98/PHF23 fusion 2 protein Nup98-Nup96 OTTHUMP00000013954; 96 kDa nucleoporin; 98 kDa nucleoporin; GLFG-repeat containing nucleoporin; Nuclear pore complex protein Nup96; Nuclear pore complex protein Nup98; Nuclear pore complex protein Nup98-Nup96; nucleoporin 98kD; nucleoporin 98kDa; Nucleoporin Nup96; Nucleoporin Nup98; Nup96; Nup98; Nup98-Nup96; NUP98/PHF23 fusion 2 protein
Gene Aliases: ADAR2; ADIR2; NUP196; NUP96; NUP98
UniProt ID: (Human) P52948
Entrez Gene ID: (Human) 4928
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.