Invitrogen
Manufacturer Code:PA554518
Catalog # PIPA554518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NUP88 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 88 kDa nucleoporin; 88 kDa nucleoporin karyoporin nuclear pore complex protein 88 nuclear pore complex protein Nup88 nucleoporin Nup88; karyoporin; nuclear pore complex protein 88; Nuclear pore complex protein Nup88; nucleoporin 88kDa; Nucleoporin Nup88
Gene Aliases: NUP88
UniProt ID: (Human) Q99567
Entrez Gene ID: (Human) 4927
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.