Invitrogen
Manufacturer Code:PA5103980
Catalog # PIPA5103980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NUP88 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 88 kDa nuclear pore complex protein 88 kDa nucleoporin karyoporin nuclear pore complex protein 88 Nuclear pore complex protein Nup88 nucleoporin 88 nucleoporin 88kDa Nucleoporin Nup84 Nucleoporin Nup88 Nup84 NUP88 Prei2 preimplantation protein 2; 88 kDa nucleoporin; karyoporin; nuclear pore complex protein 88; Nuclear pore complex protein Nup88; nucleoporin 88kDa; Nucleoporin Nup88; preimplantation protein 2
Gene Aliases: Nup84; NUP88; Prei2
UniProt ID: (Human) Q99567, (Mouse) Q8CEC0
Entrez Gene ID: (Human) 4927, (Mouse) 19069
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.