Novus Biologicals
Manufacturer Code:NBP182658
Catalog # NBP182658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QLLTRNVVFGLGGELFLWDGEDSSFLVVRLRGPSGGGEEPALSQYQRLLCINPPLFEIYQVLLSPTQHHVALIGIKGLMVLELPKRWGKNSEFEGGKSTVNCSTTPVAERFFTSSTSLTLKHAAWYPSEILDPHVV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 88 kDa nucleoporin; karyoporin; karyoporin nucleoporin 88kDa; nuclear pore complex protein 88; Nuclear pore complex protein Nup88; nucleoporin 88kDa; Nucleoporin Nup88
Gene Aliases: NUP88
UniProt ID: (Human) Q99567
Entrez Gene ID: (Human) 4927
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.