Abnova
Manufacturer Code:PAB19763
Catalog # 89-121-290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide correspoding to N-terminus ofyeast NUP84. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of NUP84.
For Research Use Only
Protein Aliases: Nuclear pore protein NUP84; Nucleoporin NUP84; Nup84p
Gene Aliases: NUP84; YDL116W
UniProt ID: (Baker's yeast) P52891
Entrez Gene ID: (Baker's yeast) 851441
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.