Novus Biologicals
Manufacturer Code:NBP29480320UL
Catalog # NB024117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 100-400 of human NUP54 (NP_059122.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 54 kDa nucleoporin; 54 kDa nucleoporin MGC13407 nucleoporin 54kD nucleoporin 54kDa nucleoporin p54; nucleoporin 54kD; nucleoporin 54kDa; Nucleoporin p54
Gene Aliases: NUP54
UniProt ID: (Human) Q7Z3B4
Entrez Gene ID: (Human) 53371
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.