Proteintech
Manufacturer Code:162321AP150UL
Catalog # 50-173-0812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NUP54 Fusion Protein Ag8882 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 54 kDa nucleoporin; 54 kDa nucleoporin nucleoporin 54kDa Nucleoporin p54 NUP54; Nuclear pore complex protein Nup54; nucleoporin 54kD; nucleoporin 54kDa; Nucleoporin Nup54; Nucleoporin p54
Gene Aliases: 3110079L04Rik; 54kDa; NUP54
UniProt ID: (Human) Q7Z3B4, (Rat) P70582, (Mouse) Q8BTS4
Entrez Gene ID: (Human) 53371, (Rat) 53372, (Mouse) 269113
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.