Invitrogen
Manufacturer Code:PA569833
Catalog # PIPA569833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat NUP50 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 50 kDa nucleoporin; nuclear pore associated protein 60 kDa; Nuclear pore complex protein Nup50; nuclear pore complex protein nup50-like protein; Nuclear pore-associated protein 60 kDa-like; nuclear pore-associated protein 60L; nucleoporin 50 kDa; Nucleoporin 50 kDa-like protein; nucleoporin 50kD; nucleoporin 50kDa; Nucleoporin Nup50; nucleoprotein 50; Nucleoprotein 50kD; NUP50 NPAP60 NPAP60L MGC39961 DKFZP564A043 nucleoporin 50kDa nuclear pore-associated protein 60L CTA-268H5.7 OTTHUMP00000199530
Gene Aliases: 1700030K07Rik; AI413123; NPAP60; NPAP60L; NUP50; PRO1146; Rtp60
UniProt ID: (Human) Q9P1K5, (Rat) O08587, (Mouse) Q9JIH2
Entrez Gene ID: (Rabbit) 100359099, (Horse) 100051494, (Dog) 474470, (Bovine) 506836, (Human) 10762, (Rat) 25497, (Mouse) 18141, (Guinea pig) 100721097
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.