Novus Biologicals
Manufacturer Code:NBP297442F
Catalog # NB102031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human NUP50 fragment. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 50 kDa nucleoporin; 50 kDa nucleoporin MGC39961 NPAP60 NPAP60LNucleoporin Nup50 nuclear pore complex protein Nup50 Nuclear pore-associated protein 60 kDa-like nuclear pore-associated protein 60L nucleoporin 50kD nucleoporin 50kDa; Nuclear pore complex protein Nup50; Nuclear pore-associated protein 60 kDa-like; nuclear pore-associated protein 60L; nucleoporin 50kD; nucleoporin 50kDa; Nucleoporin Nup50
Gene Aliases: NPAP60; NPAP60L; NUP50; PRO1146
UniProt ID: (Human) Q9P1K5
Entrez Gene ID: (Human) 10762
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.