Invitrogen
Manufacturer Code:PA571760
Catalog # PIPA571760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 289-318 amino acids from the C-terminal region of human NUP35 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 35 kDa nucleoporin; Mitotic phosphoprotein 44; MP-44; Nuclear pore complex protein Nup53; nucleoporin 35kDa; Nucleoporin NUP35; Nucleoporin NUP53; Nucleoporin NUP53 35 kDa nucleoporin Mitotic phosphoprotein 44 MP-44 Nuclear pore complex protein Nup53 Nucleoporin Nup35 NUP35 MP44 NUP53
Gene Aliases: MP-44; MP44; NP44; NUP35; NUP53
UniProt ID: (Human) Q8NFH5
Entrez Gene ID: (Human) 129401
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.