Invitrogen
Manufacturer Code:PA523372
Catalog # PIPA523372
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (294 KASTSDYQVISDRQTP 309) of NUP35 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 35 kDa nucleoporin; Mitotic phosphoprotein 44; MP-44; MP44 NP44 NUP53 35 kDa nucleoporin MP-44 mitotic phosphoprotein 44 nuclear pore complex protein Nup53 nucleoporin NUP53 nucleoporin Nup35; Nuclear pore complex protein Nup53; nucleoporin 35kDa; Nucleoporin NUP35; Nucleoporin NUP53
Gene Aliases: 2310006I24Rik; 35kDa; 5330402E05Rik; MP-44; MP44; NO44; NP44; NUP35; NUP53
UniProt ID: (Human) Q8NFH5, (Mouse) Q8R4R6
Entrez Gene ID: (Bovine) 511248, (Dog) 478830, (Human) 129401, (Mouse) 69482
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.