Invitrogen
Manufacturer Code:PA5110837
Catalog # PIPA5110837
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human NUP35 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310006I24Rik 35 kDa nucleoporin 35kDa 5330402E05Rik BOS_1614 Mitotic phosphoprotein 44 Mp44 MP-44 NO44 NP44 Nuclear pore complex protein Nup53 Nucleoporin NUP35 Nucleoporin NUP53 Nup35 Nup53; 35 kDa nucleoporin; Mitotic phosphoprotein 44; MP-44; Nuclear pore complex protein Nup53; nucleoporin 35kDa; Nucleoporin NUP35; Nucleoporin NUP53
Gene Aliases: MP-44; MP44; NP44; NUP35; NUP53
UniProt ID: (Human) Q8NFH5
Entrez Gene ID: (Human) 129401
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.