Invitrogen
Manufacturer Code:PA5114372
Catalog # PIPA5114372
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived mouse NUP214 recombinant protein (Position: E987-N1137). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 214 kDa nucleoporin; 214 kDa nucleoporin BC039282 CAIN CAN CAN protein putative oncogene D2H9S46E Kiaa0023 Nuclear pore complex protein Nup214 nucleoporin 214 nucleoporin 214kDa Nucleoporin Nup214 NUP214 Protein CAN; Nuclear pore complex protein Nup214; nucleoporin 214kDa; Nucleoporin Nup214
Gene Aliases: BC039282; CAN; D2H9S46E; Kiaa0023; Nup214; RGD1304977
UniProt ID: (Mouse) Q80U93
Entrez Gene ID: (Rat) 296634, (Mouse) 227720
Molecular Function:
RNA binding protein
nucleic acid binding
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.