Invitrogen
Manufacturer Code:PA555112
Catalog # PIPA555112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NUP205 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 205 kDa nucleoporin; Nuclear pore complex protein Nup205; nucleoporin 205kDa; Nucleoporin Nup205
Gene Aliases: 3830404O05Rik; AV248391; C7orf14; KIAA0225; mKIAA0225; NPHS13; NUP205
UniProt ID: (Human) Q92621
Entrez Gene ID: (Human) 23165, (Mouse) 70699, (Rat) 362335
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.