Invitrogen
Manufacturer Code:PA5116835
Catalog # PIPA5116835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human NUP205 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 205 kDa nucleoporin; 205 kDa nucleoporin 3830404O05Rik AV248391 C7orf14 KIAA0225 mKIAA0225 NPHS13 Nuclear pore complex protein Nup205 nucleoporin 205 nucleoporin 205kDa nucleoporin Nup205 NUP205; Nuclear pore complex protein Nup205; nucleoporin 205kDa; Nucleoporin Nup205
Gene Aliases: 3830404O05Rik; AV248391; C7orf14; KIAA0225; mKIAA0225; NPHS13; NUP205
UniProt ID: (Human) Q92621
Entrez Gene ID: (Human) 23165, (Mouse) 70699, (Rat) 362335
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.