Proteintech
Manufacturer Code:244391AP150UL
Catalog # 50-173-4361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NUP205 Fusion Protein Ag19832 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 205 kDa nucleoporin; 205 kDa nucleoporin C7orf14 KIAA0225 nucleoporin 205kDa Nucleoporin Nup205 NUP205; Nuclear pore complex protein Nup205; Nucleoporin Nup205
Gene Aliases: C7orf14; KIAA0225; NPHS13; NUP205
UniProt ID: (Human) Q92621
Entrez Gene ID: (Human) 23165
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.