Invitrogen
Manufacturer Code:PA513615
Catalog # PIPA513615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 9-38 amino acids from the N-terminal region of human NUP155 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 155 kDa nucleoporin; Nuclear pore complex protein Nup155; Nuclear pore complex protein Nup155 Nucleoporin Nup155 155 kDa nucleoporin KIAA0791 NUP155; nucleoporin 155kD; nucleoporin 155kDa; Nucleoporin Nup155
Gene Aliases: ATFB15; KIAA0791; N155; NUP155
UniProt ID: (Human) O75694
Entrez Gene ID: (Human) 9631
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.