Invitrogen
Manufacturer Code:PA5103978
Catalog # PIPA5103978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NUP155 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 155 kDa nucleoporin; 155 kDa nucleoporin ATFB15 D930027M19Rik KIAA0791 mKIAA0791 N155 Nuclear pore complex protein Nup155 nucleoporin 155 nucleoporin 155kD nucleoporin 155kDa nucleoporin Nup155 NUP155 P140; Nuclear pore complex protein Nup155; nucleoporin 155kD; nucleoporin 155kDa; Nucleoporin Nup155; P140
Gene Aliases: ATFB15; D930027M19Rik; KIAA0791; mKIAA0791; N155; NUP155
UniProt ID: (Human) O75694, (Rat) P37199, (Mouse) Q99P88
Entrez Gene ID: (Human) 9631, (Rat) 117021, (Mouse) 170762
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.