Abnova
Manufacturer Code:PAB13051
Catalog # 89-116-304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 18 amino acids of human NUP155. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NUP155.
For Research Use Only
Protein Aliases: 155 kDa nucleoporin; KIAA0791; N155; Nuclear pore complex protein Nup155; nucleoporin 155kD; nucleoporin 155kDa; Nucleoporin Nup155
Gene Aliases: ATFB15; KIAA0791; N155; NUP155
UniProt ID: (Human) O75694
Entrez Gene ID: (Human) 9631
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.