Invitrogen
Manufacturer Code:PA5115677
Catalog # PIPA5115677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Nup153 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 153 kDa nucleoporin; 153 kDa nucleoporin B130015D15Rik C88147 HNUP153 N153 nuclear pore complex protein hnup153 nuclear pore complex protein Nup153 nucleoporin 153 nucleoporin 153kD nucleoporin 153kDa nucleoporin Nup153 NUCZINK NUP153; nuclear pore complex protein hnup153; Nuclear pore complex protein Nup153; nucleoporin 153kD; nucleoporin 153kDa; Nucleoporin Nup153
Gene Aliases: B130015D15Rik; C88147; HNUP153; N153; NUCZINK; NUP153
UniProt ID: (Human) P49790
Entrez Gene ID: (Human) 9972, (Rat) 25281, (Mouse) 218210
Molecular Function: RNA binding protein nucleic acid binding structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.