Invitrogen
Manufacturer Code:PA568251
Catalog # PIPA568251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human NUP133. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 133 kDa nucleoporin; Nuclear pore complex protein Nup133; Nuclear pore complex protein Nup133 Nucleoporin Nup133 133 kDa nucleoporin NUP133; nucleoporin 133; nucleoporin 133kD; nucleoporin 133kDa; Nucleoporin Nup133
Gene Aliases: hNUP133; mermaid; NUP133
UniProt ID: (Human) Q8WUM0, (Mouse) Q8R0G9
Entrez Gene ID: (Human) 55746, (Mouse) 234865, (Rat) 292085
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.