Novus Biologicals
Manufacturer Code:NBP231795
Catalog # NBP231795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: VTQISVDLMDDYPASDPRWAESVPEEAPGFSNTSLIILHQLEDKMKAHSFLMDFIHQVGLFGRLGSFPVRGTPMATRLLLCEHAEKLSAAIVLKNHHSRLSDLVNTAILIALNKREYEIPSNLTPA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 133 kDa nucleoporin; FLJ10814 hNUP133133 kDa nucleoporin MGC21133 nuclear pore complex protein Nup133 nucleoporin 133kD nucleoporin 133kDa Nucleoporin Nup133; Nuclear pore complex protein Nup133; nucleoporin 133kD; nucleoporin 133kDa; Nucleoporin Nup133
Gene Aliases: hNUP133; NUP133
UniProt ID: (Human) Q8WUM0
Entrez Gene ID: (Human) 55746
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.