OriGene
Manufacturer Code:TA812189
Catalog # 50-168-4443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4A1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-225 of human NUMBL produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CAG3A CTG3a NBL NUMB-R NUMBLIKE NUMBR TNRC23; numb homolog (Drosophila)-like; numb homolog-like; Numb-like protein; Numb-R; Numb-related protein
Gene Aliases: CAG3A; CTG3a; NBL; NUMB-R; NUMBL; NUMBLIKE; NUMBR; TNRC23
UniProt ID: (Human) Q9Y6R0
Entrez Gene ID: (Human) 9253
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.