Proteintech
Manufacturer Code:601371IG150UL
Catalog # 50-173-6261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G12B7 |
Immunogen | peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: h Numb NUMB numb homolog (Drosophila) Protein numb homolog Protein S171 S171; h-Numb; m-Nb; numb gene homolog; numb homolog; Protein numb homolog; Protein S171
Gene Aliases: c14_5527; C14orf41; Nb; NUMB; S171
UniProt ID: (Human) P49757, (Mouse) Q9QZS3
Entrez Gene ID: (Human) 8650, (Mouse) 18222, (Rat) 29419
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.