Invitrogen
Manufacturer Code:PA578257
Catalog # PIPA578257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human NUMB. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: h-Numb; m-Nb; numb gene homolog; numb homolog; Protein numb homolog; Protein S171; S171 NUMB protein numb homolog h-Numb numb homolog (Drosophila)
Gene Aliases: c14_5527; C14orf41; Nb; NUMB; S171
UniProt ID: (Human) P49757, (Mouse) Q9QZS3
Entrez Gene ID: (Human) 8650, (Mouse) 18222, (Rat) 29419
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.