Invitrogen
Manufacturer Code:PA559680
Catalog # PIPA559680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NULP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FKSG26 Hulp1 NULP1 PRO2620 hKIAA1049 NULP1 TCF-25 nuclear localized protein 1 transcription factor 25; Nuclear localized protein 1; NULP1; TCF-25; Transcription factor 25; transcription factor 25 (basic helix-loop-helix)
Gene Aliases: FKSG26; hKIAA1049; Hulp1; KIAA1049; NULP1; PRO2620; TCF25
UniProt ID: (Human) Q9BQ70
Entrez Gene ID: (Human) 22980
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.